Anti-ATPase AAA2 domain Rabbit Polyclonal Antibody
AGRIAS111754
: Agrisera
- Antibody type:Primary
- Antigen name:ATPase AAA2 domain
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Synechococcus sp. pcc 7942,A. thaliana
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from proteins containing AAA2 domain, including Arabidopsis thaliana ClpB1 P42730, At1g74310
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: ATPase AAA2 domain
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Synechococcus sp. pcc 7942