Anti-IgA alpha Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
AGRIAS10942
: Agrisera
- Antibody type:Secondary
- Antigen symbol:IgA alpha
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human
- Cross adsorption:No
- Storage temperature:+2°C to +8 °C
- Shipping temperature:Ambient
- Immunogen:Purified Human IgA (alpha chain)
- Purification:purified
- Size:1 mg
- Pk:1 mg
Type: Secondary
Antigen: IgA alpha
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Human