Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 488)
AGRIAS10843
: Agrisera
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Flow cytometry:Yes
- Host:Donkey
- ImmunoChemistry:Yes
- Reactivity:Rabbit
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:Purified Rabbit IgG, whole molecule
- Purification:purified
- Size:1 mg
- Pk:1 mg
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Donkey
Isotype:
Reactivity: Rabbit