Anti-IgG (H+L) Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # AGRIAS10841
Supplier: Agrisera
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Host:Donkey
- ImmunoChemistry:Yes
- Reactivity:Rabbit,Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:purified rabbit IgG
- Purification:purified
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Donkey
Isotype:
Reactivity: Human, Mouse, Rabbit