Anti-Clathrin heavy-chain Rabbit Polyclonal Antibody
numer katalogowy AGRIAS10690
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:Clathrin heavy-chain
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated peptide derived from available plant clathrin heavy chain sequences including Arabidopsis thaliana Q0WNJ6, At3g11130
- Tested applications:Immunolocalization
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CLTC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human