Anti-HDEL Mouse Monoclonal Antibody [clone: 2E7]
AGRIAS10683
: Agrisera
- Antibody type:Primary
- Antigen symbol:HDEL
- Clonality:Monoclonal
- Clone:2E7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:A. thaliana
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:synthetic HDEL peptide corresponding to the C-terminus of yeast Bip
- Tested applications:Immuno-Gold
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: HDEL
Clonality: Monoclonal
Clone: 2E7
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG2b
Reactivity: A. thaliana