Anti-AtpF Rabbit Polyclonal Antibody
Catalog # AGRIAS101604
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:F0 sector of membrane-bound ATP synthase, subunit b
- Antigen symbol:ATPF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Oryza sativa
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:isolated CF0I subunit of the chloroplast ATP synthase complex
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ATPF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Oryza sativa