Anti-IgM µ Rabbit Polyclonal Antibody
Catalog # AGRIAS101506
Supplier: Agrisera
Specifications
- Antibody type:Secondary
- Antigen symbol:IgM µ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Cross adsorption:No
- Storage temperature:+2°C to +8 °C
- Shipping temperature:Ambient
- Immunogen:Purified human IgM (m chain) AAC37537
- Purification:affinity-purified
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human