Anti-IgG (H+L) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
AGRIAS101468
: Agrisera
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:Purified rabbit IgG (H&L)
- Tested applications:Dot Blot
- Purification:purified
- Size:1 mg
- Pk:1 mg
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rabbit, Rat