Order Entry
Poland
ContactUsLinkComponent
Anti-Ricin RTA subunit Mouse Monoclonal Antibody [clone: MMA]
Anti-Ricin RTA subunit Mouse Monoclonal Antibody [clone: MMA]
Catalog # AGRIAS09648
Supplier:  Agrisera
Anti-Ricin RTA subunit Mouse Monoclonal Antibody [clone: MMA]
Catalog # AGRIAS09648
Supplier:  Agrisera

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ricin RTA subunit
  • Clonality:
    Monoclonal
  • Clone:
    MMA
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Zea mays,
    A. thaliana
  • Cross adsorption:
    No
  • Storage temperature:
    –70 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    ricin, RTA subunit
  • Size:
    200 µg
  • Pk:
    200 µG

Specifications

About this item

Type: Primary
Antigen: Ricin RTA subunit
Clonality: Monoclonal
Clone: MMA
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: A. thaliana, Zea mays