Anti-BiP Goat Polyclonal Antibody
AGRIAS09615
: Agrisera
- Antibody type:Primary
- Antigen symbol:BIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Reactivity:Bovine,Rat
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from Arabidopsis thaliana BiP proteins: BiP1 At5g28540 Q9LKR3, BiP2 At5g42020 F4K007 , BiP3 At1g09080 Q8H1B3
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: BIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Rat