Anti-AtpH Rabbit Polyclonal Antibody
AGRIAS09591
: Agrisera
- Antibody type:Primary
- Antigen name:F1 sector of membrane-bound ATP synthase, delta subunit
- Antigen symbol:ATPH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Spinacia oleracea,Zea mays,A. thaliana,H. vulgare
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated peptides derived from AtpH subunit c of Arabidopsis thaliana P56760, AtCg00140 and Chlamydomonas reinhardtii Q37304
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: ATPH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, H. vulgare, Spinacia oleracea, Zea mays