Anti-IAA Rabbit Polyclonal Antibody
Catalog # AGRIAS09421
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:IAA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:A. thaliana,Medicago sativa,Euphorbia pulcherrima
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:BSA-conjugated, via C1 carobxyl group, indole-3-acetic acid (N1)
- Tested applications:Immunolocalization
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: IAA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Euphorbia pulcherrima, Medicago sativa