Anti-PTA Rabbit Polyclonal Antibody
200 µg
Catalog # AGRIAS07276
Supplier: Agrisera
Jump to:
Product Details & Documents
Type: Primary
Antigen: PTA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Pisum sativum
Documents
AS07 276
Datasheet
Specifications
- Size:200 µg
- Western blot:Yes
- Cat. No.:AGRIAS07276
- Clonality:Polyclonal
- Reactivity:Pisum sativum|A. thaliana
- Antibody type:Primary
- Immunogen:KLH-conjugated synthetic peptide conjugated derived from PTA2 of Chlamydomonas reinhardtii A8IZZ9
- Storage temperature:–20 °C
- Host:Rabbit
- Conjugation:Unconjugated
- Antigen symbol:PTA
- Cross adsorption:No
- Shipping temperature:Ambient
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



