Anti-PR-2 Rabbit Polyclonal Antibody
Catalog # AGRIAS07208
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:PR-2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Spinacia oleracea,T. aestivum,Zea mays,A. thaliana,Solanum lycopersicum,H. vulgare
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:purified tobacco class I, basic ß-1,3-glucanase. Purified GLU I consists of a mixture of closely related polypeptides encoded by a family of GLU I genes comprising GLA B5APL3 derived from the sylvestris ancestor of tobacco, GLB P27666 derived from the tomentosiformis ancestor of tobacco and homeologous recombinants (Sperisen et al., 1991). Mature GLU I is processed from a pre-pro-polypeptide (Shinshi et al., 1988).
- Tested applications:Immunolocalization
- Purification:purified
- Size:2 mg
- Pk:2 mg
Specifications
About this item
Type: Primary
Antigen: PR-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, H. vulgare, Solanum lycopersicum, Spinacia oleracea, T. aestivum, Zea mays