Anti-Cyt c6 Rabbit Polyclonal Antibody
Catalog # AGRIAS06202
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:CYT C6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:A. thaliana,Lupinus luteus
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:GST fusion of recombinant Chlamydomonas reinhardtii cytochrome c6 A8J9G6
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: CYT C6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Lupinus luteus