Anti-GS Rabbit Polyclonal Antibody
AGRIAS06183
: Agrisera
- Antibody type:Primary
- Antigen symbol:GS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Chlamydomonas reinhardtii
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from Zea mays GSH-S sequence Q8W4W2
- Tested applications:Immunolocalization
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: GS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii