Anti-PsbP Rabbit Polyclonal Antibody
Catalog # AGRIAS06167
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:PSBP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:A. thaliana
- Western blot:Yes
- Cross adsorption:No
- Molecular weight:23 kDa
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from PsbP protein of Arabidopsis thaliana Q42029, At1g06680
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PSBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana