Anti-NtrC Rabbit Polyclonal Antibody
AGRIAS06159
: Agrisera
- Antibody type:Primary
- Antigen symbol:NTRC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:A. thaliana
- Western blot:Yes
- Epitope:near the N-terminus
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated peptide derived from N-terminal domain of NtrC of Chlamydomonas reinhardtii A8HNQ7
- Size:200 µg
- Pk:200 µG
Type: Primary
Antigen: NTRC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype:
Reactivity: A. thaliana