Anti-AOX1 Rabbit Polyclonal Antibody
AGRIAS06152
: Agrisera
- Antibody type:Primary
- Antigen name:Aldehyde Oxidase 1
- Antigen symbol:AOX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:whole presumed mature AOX1 protein from from Chlamydomonas reinhardtii O65000 fused to GST
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: AOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human