Anti-FOX1 Rabbit Polyclonal Antibody
Catalog # AGRIAS06120
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:FOX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:residues 394-646 from Chlamydomonas reinhardtii FOX1 A8IZT9 fused to TrxA
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: FOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human