Anti-Lhcb4 Rabbit Polyclonal Antibody
Catalog # AGRIAS06117
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:LHCB4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Oryza sativa,Spinacia oleracea,Mesembryanthemum crystallinum,Zea mays,Pisum sativum,A. thaliana,N. tabacum,Solanum lycopersicum,Hordeum vulgare,Phaseolus vulgaris,Arachis hypogaea
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from Lhcb4 (CP29, Lhcbm4) protein sequence from Chlamydomonas reinhardtii Q93WD2
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: LHCB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Arachis hypogaea, Hordeum vulgare, Solanum lycopersicum, Mesembryanthemum crystallinum, N. tabacum, Oryza sativa, Phaseolus vulgaris, Pisum sativum, Spinacia oleracea, Zea mays