Anti-PsbE Rabbit Polyclonal Antibody
Catalog # AGRIAS06112
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:PSBE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Lotus corniculatus,Anabaena pcc 7120,H. vulgare,Phaseolus vulgaris,Oryza sativa,Ulva prolifera,Trifolium pratense,C. reinhardtii,Synechococcus sp. pcc 7942,Pisum sativum,A. thaliana,N. tabacum,T. guillardii,Ditylum brightwellii,T. pseudonana
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide chosen from PsbE protein of Arabidopsis thaliana P56779, AtCg00580
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PSBE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Ulva prolifera, Anabaena pcc 7120, A. thaliana, C. reinhardtii, Ditylum brightwellii, H. vulgare, Lotus corniculatus, N. tabacum, Oryza sativa, Phaseolus vulgaris, Pisum sativum, Synechococcus sp. pcc 7942, T. guillardii, T. pseudonana, Trifolium pratense