Anti-IKZF3 Mouse monoclonal Antibody (Biotin) [clone: 5H2]
Catalog # ORIGTA700525
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:IKZF3
- Clonality:Monoclonal
- Clone:5H2
- Conjugation:Biotin
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Epitope:Full length
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Stored in PBS (pH 7,4) with 0,05% sodium azide, 10mg/ml BSA, 50% glycerol
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human IKZF3 (NM_012481) produced in HEK293T cell.
- Size:50 μg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: IKZF3
Clonality: monoclonal
Clone: 5H2
Conjugation: Biotin
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human