Anti-FKBP4 Mouse Monoclonal Antibody [clone: Hi52C]
ORIGTA326427
: OriGene
- Antibody type:Primary
- Antigen symbol:FKBP4
- Clonality:Monoclonal
- Clone:Hi52C
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Hamster,Human,Dog,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS, 50% glycerol, 0,09% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Synthetic peptide corresponding to the residues of human FKBP52
- Purification:purified by Protein G chromatography
- Size:100µg
- Pk:100 µG
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 5?g with 10-20?l Protein A beads; Immunochemistry: 1:250
Type: Primary
Antigen: FKBP4
Clonality: Monoclonal
Clone: Hi52C
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: canine, Hamster, Human, Mouse, Rat