Order Entry
Poland
ContactUsLinkComponent
 
Anti-ASPSCR1 Rabbit Polyclonal Antibody
  ORIGTA324202
 :  OriGene
Anti-ASPSCR1 Rabbit Polyclonal Antibody
  ORIGTA324202
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen name:
    alveolar soft part sarcoma chromosome region, candidate 1
  • Antigen symbol:
    ASPSCR1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Amino acid number:
    303 to 307
  • Storage buffer:
    PBS pH 7,3, 0,05% NaN3, 50% glycerol
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Peptide sequence around aa.303 ~307(P-Q-Q-E-Q) derived from Human ASPSCR1.
  • Purification:
    purified by antigen affinity chromatography
  • Size:
    100 μl
  • Pk:
    100 µl

 

 

Recommended Dilutions: Western Blot: 1:500-1000

Type: Primary
Antigen: ASPSCR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human