Anti-RARA Rabbit Polyclonal Antibody
ORIGTA324529
: OriGene
- Antibody type:Primary
- Antigen name:Retinoic Acid Receptor Alpha
- Antigen symbol:RARA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Liquid
- Amino acid number:322 to 349
- Storage buffer:Purified polyclonal antibody supplied in PBS with 0,09% (W/V) sodium azide.
- Molecular weight:50.77 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:This RARA antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 322-349 amino acids from the C-terminal region of human RARA.
- Purification:purified by Protein chromatography, followed by peptide affinity purification
- Size:400 μl
- Pk:400 µl
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:10-50; Immunochemistry: 1:10-50; Flow Cytometry: 1:10-50
Type: Primary
Antigen: RARA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse