Anti-ITGB3 Mouse Polyclonal Antibody [clone: VI-PL2]
Catalog # ORIGTA320397
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:ITGB3
- Clonality:Polyclonal
- Clone:VI-PL2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Baboon,Rhesus monkey,Human,Dog
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Aqueous buffer, 0,09% sodium azide, may contain carrier protein/stabilizer
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Tested applications:Functional Assay
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ITGB3
Clonality: Polyclonal
Clone: VI-PL2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Baboon, canine, Human, Rhesus