Anti-BCL2L11 Mouse Monoclonal Antibody [clone: 1C2H4]
ORIGTA319806
: OriGene
- Antibody type:Primary
- Antigen symbol:BCL2L11
- Clonality:Monoclonal
- Clone:1C2H4
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:2 to 198
- Storage buffer:Bim Monoclonal Antibody is supplied in PBS containing 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Mouse monoclonal Bim antibody was raised against a recombinant protein corresponding to amino acids 2 to 198 of human Bim.
- Purification:purified by immunoaffinity chromatography
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunochemistry: 10 ?g/ml; Immunofluorescence: 20 ?g/ml
Type: Primary
Antigen: BCL2L11
Clonality: Monoclonal
Clone: 1C2H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat