Order Entry
Poland
ContactUsLinkComponent
Anti-CD55 Mouse Polyclonal Antibody [clone: 143-30]
Anti-CD55 Mouse Polyclonal Antibody [clone: 143-30]
Catalog # ORIGTA320286
Supplier:  OriGene
Anti-CD55 Mouse Polyclonal Antibody [clone: 143-30]
Catalog # ORIGTA320286
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD55
  • Clonality:
    Polyclonal
  • Clone:
    143-30
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    Aqueous buffer, 0,09% sodium azide, may contain carrier protein/stabilizer
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: CD55
Clonality: Polyclonal
Clone: 143-30
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human