Order Entry
Poland
ContactUsLinkComponent
Anti-STAT5A Mouse Monoclonal Antibody [clone: 5F6.F1]
Anti-STAT5A Mouse Monoclonal Antibody [clone: 5F6.F1]
Catalog # ORIGTA319572
Supplier:  OriGene
Anti-STAT5A Mouse Monoclonal Antibody [clone: 5F6.F1]
Catalog # ORIGTA319572
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    STAT5A
  • Clonality:
    Monoclonal
  • Clone:
    5F6.F1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Modification:
    phospho-Tyr694
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Anti-STAT5 phospho Y694 monoclonal antibody was produced by repeated immunizations with a synthetic peptide corresponding to residues surrounding Y694 of mouse STAT5a protein.
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:500-1:2000; Immunochemistry: 20-40 ?g/ml; Immunofluorescence: 1:50-1:1000

Type: Primary
Antigen: STAT5A
Clonality: Monoclonal
Clone: 5F6.F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat