Order Entry
Poland
ContactUsLinkComponent
 
Anti-TRIM9 Mouse Monoclonal Antibody [clone: 1E4]
Catalog # ORIGCF800073
Supplier:  OriGene
Anti-TRIM9 Mouse Monoclonal Antibody [clone: 1E4]
Catalog # ORIGCF800073
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    TRIM9
  • Clonality:
    Monoclonal
  • Clone:
    1E4
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Amino acid number:
    284 to 669
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    61.2 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Human recombinant protein fragment corresponding to amino acids 284-669 of human TRIM9 (NP_443210) produced in E.coli.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TRIM9
Clonality: Monoclonal
Clone: 1E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human