Anti-NUDT6 Mouse Monoclonal Antibody [clone: 9F5]
Catalog # ORIGCF502330
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:NUDT6
- Clonality:Monoclonal
- Clone:9F5
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:35.5 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human NUDT6 (NP_009014) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100
Type: Primary
Antigen: NUDT6
Clonality: Monoclonal
Clone: 9F5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human