Anti-PLEK Mouse Monoclonal Antibody [clone: 4F10]
Catalog # ORIGCF500899
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:PLEK
- Clonality:Monoclonal
- Clone:4F10
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Epitope:full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:40.1 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human PLEK (NP_002655) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:100
Type: Primary
Antigen: PLEK
Clonality: Monoclonal
Clone: 4F10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human