Anti-SQSTM1 Mouse Monoclonal Antibody [clone: 5A2]
Catalog # ORIGCF502131
Supplier: OriGene
Specifications
- Typ przeciwciała:Primary
- Symbol antygenu:SQSTM1
- Klonalność:Monoklonalne
- Klonuj:5A2
- Koniugacja:Unconjugated
- Cytometrii przepływowej:Yes
- Gospodarz:Mouse
- ImmunoChemia:Yes
- ImmunoFluorescencja:Yes
- Izotyp:IgG1
- Reaktywność:Monkey,Human
- Western blot:Yes
- Epitop:Full length
- Adsorpcja krzyżowa:No
- Formularz:Lyophilized
- Bufor do przechowywania:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Waga molekularna:47.5 kDa
- Temperatura przechowywania:–20 °C
- Temperatura transportu:–20 °C
- Immunogen:Full length human recombinant protein of human SQSTM1 (NP_003891) produced in HEK293T cell.
- Oczyszczenie:purified by affinity chromatography
- Rozmiar:100 μg
- Ilość:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: SQSTM1
Clonality: Monoclonal
Clone: 5A2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey