Anti-PARP1 Mouse Monoclonal Antibody [clone: 7A10]
ORIGTA318859
: OriGene
- Antibody type:Primary
- Antigen symbol:PARP1
- Clonality:Monoclonal
- Clone:7A10
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Supplied in 10 mM HEPES sodium (pH 7,5), 150 mM NaCl, 100 µg/ml BSA, 50% glycerol, and 0,03% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to residues before the cleavage site of human PARP-1 was used as immunogen. The antibody only recognizes full-length form (p116) of PARP-1.
- Size:100 μl
- Pk:100 µG
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:200-400
Type: Primary
Antigen: PARP1
Clonality: Monoclonal
Clone: 7A10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype:
Reactivity: Human