Anti-IgG Goat Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # BSBTBA1090-0.5
Supplier: Boster Bio
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Conjugation:TRITC (Tetramethylrhodamine Isothiocyanate)
- Host:Goat
- Cross adsorption:No
- Size:500 μg
- Pk:0,5 mg
Specifications
About this item
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanante)
Epitope:
Host: Goat
Isotype:
Reactivity: