Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 4 (anion exchanger), member 1 (Diego blood group)
- Antigen symbol:Solute carrier family 4, member 1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Gene ID:6521
- Antigen synonyms:BND3|RTA1A|FR|WD1|CD233|EPB3|EMPB3|SW|DI|WD|AE1|WR
- UniProtKB:P02730
- Storage temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Immunogen:E. coli-derived human Band 3 (Position: E28-N365). Human Band 3 shares 75.7% and 74.5% amino acid (aa) sequence identity with mouse and rat Band 3, respectively.
- Purification:Immunogen affinity purified.
- Size:100 µg/vial
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Band 3 anion transport protein(SLC4A1) detection. Tested with WB, IHC-P, IHC-F in Human;Mouse;Rat.
Band 3 is also known as SLC4A1. The protein encoded by this gene is part of the anion exchanger (AE) family and is expressed in the erythrocyte plasma membrane, where it functions as a chloride/bicarbonate exchanger involved in carbon dioxide transport from tissues to lungs. The protein comprises two domains that are structurally and functionally distinct. The N-terminal 40kDa domain is located in the cytoplasm and acts as an attachment site for the red cell skeleton by binding ankyrin. The glycosylated C-terminal membrane-associated domain contains 12-14 membrane spanning segments and carries out the stilbene disulphonate-sensitive exchange transport of anions. The cytoplasmic tail at the extreme C-terminus of the membrane domain binds carbonic anhydrase II. The encoded protein associates with the red cell membrane protein glycophorin A and this association promotes the correct folding and translocation of the exchanger. This protein is predominantly dimeric but forms tetramers in the presence of ankyrin. Many mutations in this gene are known in man, and these mutations can lead to two types of disease: destabilization of red cell membrane leading to hereditary spherocytosis, and defective kidney acid secretion leading to distal renal tubular acidosis. Other mutations that do not give rise to disease result in novel blood group antigens, which form the Diego blood group system.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: SLC6A4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat