Anti-NAD+-dependent 15-hydroxy PGDH Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Catalog # COBSD5-1864
Supplier: Columbia Biosciences
Specifications
- Antibody type:Primary
- Antigen name:NAD+-dependent 15-hydroxy PGDH
- Clonality:Polyclonal
- Conjugation:RPE (R-Phycoerythrin)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Cross adsorption:none
- Amino acid number:92 to 105
- Sequence:AGVNNEKNWEKTLQ
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: NAD+-dependent 15-hydroxy PGDH
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: