Order Entry
Poland
ContactUsLinkComponent
 
Anti-HA tag Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 16B12]
  COBSD5-1722
Anti-HA tag Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 16B12]
  COBSD5-1722

 

  • Antibody type:
    Tag
  • Antigen name:
    HA tag
  • Antigen symbol:
    HA
  • Clonality:
    Monoclonal
  • Clone:
    16B12
  • Conjugation:
    RPE (R-Phycoerythrin)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Tag
Antigen: HA tag (HA tag)
Clonality: monoclonal
Clone: 16B12
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: