Order Entry
Poland
ContactUsLinkComponent
 
Anti-schistosomal Glutathione-S-transferase Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
  COBSD5-1310
Anti-schistosomal Glutathione-S-transferase Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
  COBSD5-1310

 

  • Antibody type:
    Secondary
  • Antigen name:
    schistosomal Glutathione-S-transferase
  • Antigen symbol:
    schistosomal Glutathione-S-Transferase
  • Clonality:
    Polyclonal
  • Conjugation:
    RPE (R-Phycoerythrin)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Secondary
Antigen: schistosomal Glutathione-S-Transferase
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: