Anti-YAP1 Mouse Monoclonal Antibody [clone: 2F12]
Catalog # USBIY1200-01H
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Yes-associated Protein 1
- Antigen symbol:YAP1
- Clonality:Monoclonal
- Clone:2F12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:53 to 162
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: YAP1 (Yes-associated protein 1)
Clonality: monoclonal
Clone: 2F12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human