Order Entry
Poland
ContactUsLinkComponent
Anti-VIM Mouse Monoclonal Antibody [clone: VI-10]
Anti-VIM Mouse Monoclonal Antibody [clone: VI-10]
Catalog # USBIV2122-02Q
Supplier:  US Biological
Anti-VIM Mouse Monoclonal Antibody [clone: VI-10]
Catalog # USBIV2122-02Q
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Vimentin
  • Antigen symbol:
    VIM
  • Clonality:
    Monoclonal
  • Clone:
    VI-10
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 15mM sodium azide, pH 7,4.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by size exclusion chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: VIM (vimentin)
Clonality: monoclonal
Clone: VI-10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human