Anti-VIM Mouse Monoclonal Antibody [clone: VI-RE/1]
USBIV2122-01R
- Antibody type:Primary
- Antigen name:Vimentin
- Antigen symbol:VIM
- Clonality:Monoclonal
- Clone:VI-RE/1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 15mM sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: VIM (vimentin)
Clonality: monoclonal
Clone: VI-RE/1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human