Anti-Ubiquitin-activating Enzyme E1L Rabbit Polyclonal Antibody
Catalog # USBIU1003-01B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin-activating Enzyme E1L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,01% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 10 ?g/ml; Western Blot: 1:500-1:2000; ELISA: 1:15000-1:75000
Type: Primary
Antigen: Ubiquitin-activating Enzyme E1L
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human