Anti-TUBB3 Mouse Monoclonal Antibody [clone: TU-20]
Catalog # USBIT9155-03N
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:tubulin, beta 3 class III
- Antigen symbol:TUBB3
- Clonality:Monoclonal
- Clone:TU-20
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Amino acid number:441 to 448c
- Storage buffer:PBS, 15mM sodium azide, pH 7,4.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: TUBB3 (tubulin, beta 3 class III)
Clonality: monoclonal
Clone: TU-20
Conjugation: unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human