Anti-TUBB3 Mouse Monoclonal Antibody [clone: TU-20]
USBIT9155-03J
- Antibody type:Primary
- Antigen name:tubulin, beta 3 class III
- Antigen symbol:TUBB3
- Clonality:Monoclonal
- Clone:TU-20
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Hamster,Pig,Monkey,Bovine,Rat,Mouse
- Western blot:Yes
- Format:ascites
- Form:liquid
- Storage buffer:PBS, 15mM sodium azide, pH 7,4.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: TUBB3 (tubulin, beta 3 class III)
Clonality: monoclonal
Clone: TU-20
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Hamster, Monkey, Mouse, Pig, Rat