Anti-Thyroxine Sheep Polyclonal Antibody
Catalog # USBIT5460-01K
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Thyroxine
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Isotype:IgG
- Format:Serum
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Radio Immunoassay
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Recommended dilution: Radio Immunoassay: 1:19
Type: Primary
Antigen: Thyroxine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: