Order Entry
Poland
ContactUsLinkComponent
Anti-Thyroxin Rabbit Polyclonal Antibody
Anti-Thyroxin Rabbit Polyclonal Antibody
numer katalogowy USBIT5460-01J-100
Dostawca:  US Biological
Anti-Thyroxin Rabbit Polyclonal Antibody
numer katalogowy USBIT5460-01J-100
Dostawca:  US Biological

Dane techniczne

  • Antibody type:
    Primary
  • Antigen symbol:
    Thyroxin
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Format:
    Serum
  • Form:
    lyophilized powder
  • Storage buffer:
    50mM Tris, pH 7,2.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Tested applications:
    Radio Immunoassay
  • Size:
    100 µl
  • Pk:
    100 µl

Dane techniczne

Informacje o tym produkcie

Recommended dilution: Radio Immunoassay: 1:1.000

Type: Primary
Antigen: Thyroxin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: