Anti-TCR, Cbeta1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 7H199]
Catalog # USBIT2040-04A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:TCR C beta1
- Clonality:Monoclonal
- Clone:7H199
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,5, 0,2% BSA, 0,04% sodium azide,5% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:120 Tests
- Pk:120 Tests
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:50
Type: Primary
Antigen: TCR, Cbeta1
Clonality: monoclonal
Clone: 7H199
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human